Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPY Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130929

Catalog No. PIPA130929


Add to cart

Description

Description

PA1-30929 detects NPY Y2 from rat, mouse samples. PA1-30929 has been successfully used in immunohistochemistry applications. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

NPY
Polyclonal
PBS with 0.05% sodium azide
P57774, P07808
NPY
Proprietary immunogen - Contact ABR for details.
Affinity chromatography
RUO
Antibody
Polyclonal
Mouse, Rat
Immunohistochemistry
Unconjugated
Npy
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
109648, 24604
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit