Learn More
MilliporeSigma™ anti-NQO1, Polyclonal Available on GSA/VA Contract for Federal Government customers only.
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ ST1535100UG
Catalog No. ST1535100UG
Add to cart
Description
Description
Specifically detects NQO1 Clone: in Human, Mouse samples, and it is validated for Immuno Blotting
NQO1 is a member of the NAD(P)H dehydrogenase (quinone) family and encodes a cytoplasmic 2-electron reductase. This FAD-binding protein forms homodimers and reduces quinones to hydroquinones. This protein's enzymatic activity prevents the one electron reduction of quinones that results in the production of radical species. Mutations in this gene have been associated with tardive dyskinesia (TD), an increased risk of hematotoxicity after exposure to benzene, and susceptibility to various forms of cancer. Altered expression of this protein has been seen in many tumors and is also associated with Alzheimer's disease (AD).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
NQO1
Unconjugated
Purified
Full-length, recombinant human NQO1 (aa 1-274)
100μg
−20°C
Polyclonal
Human, Murine
Immuno Blotting, Immunoblot
In PBS, pH 7.2.
Rabbit
IgG
RUO
Primary
Recognizes the ∽30kDa NQO1 protein in human stomach tissue, mouse brain tissue, and HepG2 cells.