Learn More
NQO2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA528328
Catalog No. PIPA528328
Add to cart
Description
Description
PA5-28330 targets CSEN in WB applications and shows reactivity with Human and mouse samples. The PA5-28330 immunogen is recombit fragment corresponding to a region within amino acids 1 and 256 of Human CSEN.
NQO2 (EC 1.10.99.2) is a flavoprotein that catalyzes the 2-electron reduction of various quinones, redox dyes, and the vitamin K menadione. NQO2 predomitly uses dihydronicotinamide riboside (NRH) as the electron donor (summary by Wu et al., 1997 [PubMed 9367528]).
Specifications
Specifications
NQO2
Polyclonal
Unconjugated
NQO2
Liquid
NQO2
Recombit fragment corresponding to a region within amino acids 1 and 231 of Human NQO2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P16083, Q9JI75
DHQV, DIA6, NMOR2, QR2, NAD(P)H menadione oxidoreductase 2, dioxin-inducible, NAD(P)H menadione oxidoreductase-1, dioxin-inducible-2, NRH dehydrogenase [quinone] 2, NRH:quinone oxidoreductase 2, quinone reductase 2, ribosyldihydronicotinamide dehydrogenase, ribosyldihydronicotinamide
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18105, 4835