Learn More
NR1 Mouse anti-Mouse, Rat, AlexaFluor™488, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05432A4
Catalog No. 05-432-A4MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect NR1 using this Anti-NR1 Antibody, C Terminus, Alexa Fluor™488 Conjugate validated for use in immunocytochemistry, immunohistochemistry.
Specifications
Specifications
NR1
Alexa Fluor 488
NP_058706
Grin1, Nmdar1
Recombinant protein corresponding to rat NR1.
Protein G Purified
2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Monoclonal
Immunocytochemistry, Immunohistochemistry
Purified mouse monoclonal IgG conjugated to Alexa Fluor™ 488 in PBS with 0.1% sodium azide and 15mg/mL BSA.
Purified
Murine
IgG
100μL
Primary
Murine, Rat