Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LXR alpha Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526951

Catalog No. PIPA526951

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

NR1H3 form a subfamily of the nuclear receptor superfamily and are key regulators of macrophage function, controlling transcriptional programs involved in lipid homeostasis and inflammation. The inducible LXRA is highly expressed in liver, adrenal gland, intestine, adipose tissue, macrophages, lung, and kidney, whereas LXRB is ubiquitously expressed. Ligand-activated LXRs form obligate heterodimers with retinoid X receptors (RXRs; see MIM 180245) and regulate expression of target genes containing LXR response elements.
Specifications

Specifications

LXR alpha
Polyclonal
PBS with 0.09% sodium azide
Q13133
LXR-a, LXRA, RLD-1, liver X nuclear receptor alpha variant 1, oxysterols receptor LXR-alpha
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10062
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
NR1H3
Liquid
NR1H3
KLH conjugated synthetic peptide between 226-253 amino acids from the central region of human NR1H3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit