Learn More
NR2A Rabbit anti-Mouse, Rat, Clone: A12W, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier:  MilliporeSigma 04901
Catalog No. 04-901-MI
Add to Cart
Description
Description
Anti-NR2A Antibody, clone A12W is an antibody against NR2A for use in western blotting.
Specifications
Specifications
NR2A
Monoclonal
Unconjugated
NM_008170
6His-tagged fusion protein corresponding to amino acids 1265-1464 of mouse NMDA receptor 2A (NR2A)
Primary
Mouse, Rat
Supernatant
Western Blot
A12W
Cultured supernantant in 0.05% sodium azide
Rabbit
100 μL
GRIN2A, hNR2A, Glutamate, NMDAR2A, NR2A
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
IgG
Recommendations
Recommendations