Learn More
NR2A Rabbit anti-Rat, Human, Mouse, Clone: A12W, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05901R
Catalog No. 05-901-RMI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This Anti-NR2A Antibody, clone A12W is validated for use in western blotting for the detection of NR2A.
Specifications
Specifications
NR2A
A12W
Purified Rabbit Monoclonal IgG in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide and 40% Glycerol.
Purified
Histidine-tagged recombinant protein corresponding to the C-terminus of mouse NR2A.
Protein A
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Monoclonal
Human, Murine, Rat
Western Blot
Unconjugated
NP_032196
Rabbit
IgG
100μL
Primary
GRIN2A, hNR2A, Glutamate, NMDAR2A, NR2A