Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR2C2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human NR2C2 protein.

Manufacturer:  Abnova Corporation H00007182D01P

Catalog No. 89-002-961


Add to cart

Description

Description

Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes (Yoshikawa et al., 1996 [PubMed 8661150]).[supplied by OMIM

Sequence: MTSPSPRIQIISTDSAVASPQRIQGSEPASGPLSVFTSLNKEKIVTDQQTGQKIQIVTAVDASGSPKQQFTLTSPDGAGTGKVILASPETSSAKQLIFTTSDNLVPGRIQIVTDSASVERLLGKTDVQRPQVVEYCVVCGDKASGRHYGAVSCEGCKGFFKRSVRKNLTYSCRSNQDCIINKHHRNRCQFCRLKKCLEMGMKMESVQSKRKPFDVQREKPSNCAASTEKIYIRKDLRSPLIATPTFVADKDGARQTGLLDPGMLVNIQQPLIREDGTVLLATDSKAETSQGALGTLANVVTSLANLSESLNNGDTSEIQPEDQSASEITRAFDTLAKALNTTDSSSSPSLADGIDTSGGGSIHVISRDQSTPIIEVEGPLLSDTHVTFKLTMPSPMPEYLNVHYICESASRLLFLSMHWARSIPAFQALGQDCNTSLVRACWNELFTLGLAQCAQVMSLSTILAAIVNHLQNSIQEDKLSGDRIKQVMEHIWKLQEFCNSMAKLDIDGYEYAYLKAIVLFSPGKICGGDA
Specifications

Specifications

NR2C2
Unconjugated
In 1X PBS, pH 7.4
BC051670.1
TAK1|TR2R1|TR4|hTAK1
Rabbit
100 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Western Blot
Rabbit polyclonal antibody raised against a full-length human NR2C2 protein.
NR2C2
Purified
NR2C2
NR2C2 (AAH51670.1, 1 a.a. ~ 530 a.a) full-length human protein.
RUO
Primary
7182
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit