Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR2E1, Mouse, Clone: 1C4, Abnova™

Mouse monoclonal antibody raised against a partial recombinant NR2E1.

Manufacturer:  Abnova Corporation H00007101M01

Catalog No. 89-002-705


Add to cart

Description

Description

Sequence: NGDNTDSQKLNKIISEIQALQEVVARFRQLRLDATEFACLKCIVTFKAVPTHSGSELRSFRNAAAIAALQDEAQLTLNSYIHTRYPTQPCRFGK
Specifications

Specifications

NR2E1
1C4
Mouse monoclonal antibody raised against a partial recombinant NR2E1.
NR2E1
Liquid
NR2E1
NR2E1 (NP_003260, 249 a.a. ~ 342 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Murine
ELISA, Western Blot
Unconjugated
In 1X PBS, pH 7.4
NM_003269
TLL|TLX|XTLL
Mouse
IgG2a Kappa
RUO
Primary
7101
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit