Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR2F2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528033

Catalog No. PIPA528033


Add to cart

Description

Description

PA5-28033 targets NR2F2 in WB applications and shows reactivity with Human and mouse samples. The PA5-28033 immunogen is recombit fragment corresponding to a region within amino acids 123 and 383 of Human NR2F2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a member of the steroid thyroid hormone superfamily of nuclear receptors. The encoded protein is a ligand inducible transcription factor that is involved in the regulation of many different genes. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

NR2F2
Polyclonal
Unconjugated
NR2F2
Liquid
NR2F2
Recombit fragment corresponding to a region within amino acids 123 and 383 of Human NR2F2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.57 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P24468, P43135
ARP1, COUPTFB, COUPTFII, NF-E3, NR2F1, SVP40, TFCOUP2, ADP-ribosylation factor related protein 1, COUP transcription factor 2, COUP transcription factor II, apolipoprotein A-I regulatory protein 1, apolipoprotein AI regulatory protein 1, chicken ovalbumin upstream promoter transcription factor 2, chicken ovalbumin
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11819, 7026
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit