Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucocorticoid Receptor Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524793
Description
NR3C1 is a receptor for glucocorticoids that can act as both a transcription factor and as a regulator of other transcription factors. This protein can also be found in heteromeric cytoplasmic complexes along with heat shock factors and immunophilins. The protein is typically found in the cytoplasm until it binds a ligand, which induces transport into the nucleus. Mutations in this gene are a cause of glucocorticoid resistance, or cortisol, resistance.Specifications
Glucocorticoid Receptor | |
Polyclonal | |
NR3C1 | |
GCCR, GCR, GR, GRL, glucocorticoid nuclear receptor variant 1, glucocorticoid receptor, nuclear receptor subfamily 3 group C member 1 variant DL-1, nuclear receptor subfamily 3 group C member 1 variant NS-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2908 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
P04150 | |
NR3C1 | |
KLH conjugated synthetic peptide between 713-742 amino acids from the C-terminal region of human NR3C1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction