Learn More
NR5A1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525030
Catalog No. PIPA525030
Add to cart
Description
Description
This antibody is predicted to react with bovine, chicken and rat based on sequence homology.
NR5A1 is a transcriptional activator involved in sex determination. The encoded protein binds DNA as a monomer. Defects in this protein are a cause of XY sex reversal with or without adrenal failure as well as adrenocortical insufficiency without ovarian defect.
Specifications
Specifications
NR5A1
Polyclonal
Unconjugated
NR5A1
Liquid
NR5A1
KLH conjugated synthetic peptide between 64-93 amino acids from the N-terminal region of human NR5A1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q13285
AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1, SPGF8, SRXY3, STF-1, adrenal 4 binding protein, adrenal 4-binding protein, fushi tarazu factor homolog 1, nuclear receptor AdBP4, steroid hormone receptor Ad4BP, steroidogenic factor 1, steroidogenic factor 1 nuclear receptor, steroidogenic factor-1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2516