Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219581
Description
NRAS Polyclonal specifically detects NRAS in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
NRAS | |
Polyclonal | |
Unconjugated | |
PBS, 20% Glycerol (pH7). with 0.025% Proclin 300 | |
ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog | |
Rabbit | |
21 kDa | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, mTOR Pathway | |
4893 | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
0.92mg/mL | |
Western Blot 1:500-1:3000 | |
P01111 | |
NRAS | |
Full length human N-Ras Recombinant protein. | |
Affinity Purified | |
RUO | |
Primary | |
Mouse. Species reactivity based on sequence identity: Human (100%),Bovine (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only