Learn More
MilliporeSigma™ anti-NRG1, Polyclonal Available on GSA/VA Contract for Federal Government customers only.
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ ST1576100UG
Catalog No. ST1576100UG
Add to cart
Description
Description
Specifically detects NRG1 Clone: in Human samples, and it is validated for Immuno Blotting
The protein encoded by this gene was originally identified as a 44-kD glycoprotein that interacts with the NEU/ERBB2 receptor tyrosine kinase to increase its phosphorylation on tyrosine residues. This protein is a signaling protein that mediates cell-cell interactions and plays critical roles in the growth and development of multiple organ systems. The gene dysregulation has been linked to diseases such as cancer, schizophrenia and bipolar disorder (BPD).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
NRG1
Unconjugated
Purified
Full-length, human NRG1 (aa 1-241)
100μg
−20°C
Polyclonal
Human
Immuno Blotting, Immunoblot
In PBS, pH 7.2.
Rabbit
IgG
RUO
Primary
Recognizes the ∽85-90kDa NRG1 protein in human placenta tissue.