Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus Type 2 NS1 Rabbit anti-Virus, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527885

Catalog No. PIPA527885


Add to cart

Description

Description

PA5-27885 targets NS1 (Dengue virus 2) in IF and WB applications and shows reactivity with Virus samples. The PA5-27885 immunogen is synthetic peptide corresponding to a region within amino acids 51 and 144 of Dengue virus Non-structural protein 1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Dengue fever and dengue hemorrhagic fever are acute febrile diseases, found in the tropics and Africa. Caused by one of four closely related virus serotypes of the genus Flavivirus, family Flaviviridae, each serotype is sufficiently different that there is no cross-protection and epidemics caused by multiple serotypes (hyperendemicity) can occur. Dengue is transmitted to humans by the Aedes aegypti (rarely Aedes albopictus) mosquito, which feeds during the day.
Specifications

Specifications

Dengue Virus Type 2 NS1
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Virus
Immunocytochemistry, Immunofluorescence, Western Blot
1.39 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Dengue NS1; Dengue NS1 protein
Synthetic peptide corresponding to a region within amino acids 51 and 144 of NS1 (Dengue virus 2 )
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit