Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue Virus Type 2 NS5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527888
Description
Recommended positive controls: Dengue virus type 2 infected BHK-21, Dengue virus type 3 infected BHK-21. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Dengue Virus Type 2 NS5 Polyclonal antibody specifically detects Dengue Virus Type 2 NS5 in Virus samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Dengue Virus Type 2 NS5 | |
Polyclonal | |
Unconjugated | |
Dengue NS5; Dengue NS5 protein | |
Synthetic peptide corresponding to a region within amino acids 829 and 900 of NS5 (Dengue virus 2 ) | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1.59 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction