Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR74 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520925
Description
A suggested positive control is rat liver tissue lysate. PA5-20925 can be used with blocking peptide PEP-1039.
WDR74 Polyclonal antibody specifically detects WDR74 in Human, Rat samples. It is validated for Western Blot.Specifications
WDR74 | |
Polyclonal | |
Unconjugated | |
WDR74 | |
5730436H21Rik; AA407588; NOP seven-associated protein 1; NSA1; WD repeat domain 74; WD repeat-containing protein 74; Wdr74 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54663, 690229 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6RFH5 | |
WDR74 | |
An 18 amino acid peptide near the amino terminus of human NSA1. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction