Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-NSD3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP104938SS

 View more versions of this product

Catalog No. NBP104938SS


Add to cart

Description

Description

NSD3 Polyclonal antibody specifically detects NSD3 in Human samples. It is validated for Western Blot, ICC/IF (Negative).
Specifications

Specifications

NSD3
1 mg/mL
Western Blot 1 ug/ml, ICC/IF (Negative)
Q9BZ95
EC 2.1.1.43, FLJ20353, histone-lysine N-methyltransferase NSD3, MGC126766, MGC142029, NSD3DKFZp667H044, Nuclear SET domain-containing protein 3, pp14328, Protein whistle, WHSC1-like 1 isoform 9 with methyltransferase activity to lysine, WHSC1-like protein 1, Wolf-Hirschhorn syndrome candidate 1-like 1, Wolf-Hirschhorn syndrome candidate 1-like protein 1
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
This has been observed to recognize a band that corresponds to isoform 2 of NSD3.
Western Blot, Immunocytochemistry
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
Affinity Purified
WHSC1L1
Synthetic peptide made to an internal portion of the human NSD3 protein (within residues 200-300). [Swiss-Prot# Q9BZ95]
156 kDa
0.025 ml
Cancer
Primary
54904
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only