Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-NSD3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $314.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Synthetic peptide made to an internal portion of the human NSD3 protein (within residues 200-300). [Swiss-Prot# Q9BZ95]
Antigen NSD3
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP104938SS
View Documents
Novus Biologicals
NBP104938SS
0.025 ml
Each for $119.60
Add to cart
 
NBP104938
View Documents
Novus Biologicals
NBP104938
0.1 ml
Each for $314.00
Add to cart
 
Description

Description

NSD3 Polyclonal antibody specifically detects NSD3 in Human samples. It is validated for Western Blot, ICC/IF (Negative).
Specifications

Specifications

Rabbit
Synthetic peptide made to an internal portion of the human NSD3 protein (within residues 200-300). [Swiss-Prot# Q9BZ95]
Polyclonal
Primary
Western Blot, Immunocytochemistry
Q9BZ95
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
IgG
NSD3
Affinity Purified
RUO
This has been observed to recognize a band that corresponds to isoform 2 of NSD3.
54904
1 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit