Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSF Mouse, Unlabeled, Clone: 7, BD

Mouse Monoclonal Antibody

$213.00

Specifications

Antigen NSF
Isotype IgG2b
Immunogen Human NSF aa. 488-601
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB612272
View Documents Promotion Details
BD Biosciences
612272
50μg
Each for $213.00
Add to cart
 
BDB612273
View Documents Promotion Details
BD Biosciences
612273
150μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Eukaryotic protein trafficking involves the packaging of target molecules into membraneous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their components into an acceptor compartment. Components of both the vesicle and the plasma membrane interact to form a fusion complex, which mediates specific docking and fusion of vesicles. This complex contains NSF (N-ethyl-maleimide-sensitive factor), SNAPs (soluble NSF attachment proteins), and receptor proteins (SNAREs). SNAP-25 (synaptosome-associated protein of 25kDa) and syntaxin are SNAREs that are associated with the target plasma membrane (t-SNAREs), while synaptobrevin and synaptotagmin are vesicle-associated SNARE proteins (v-SNAREs). SNAREs form a 7S complex that has a high affinity binding site for α-SNAP and NSF. Upon binding of α-SNAP and NSF, a 20S complex is formed that is rapidly disassembled due to NSF's ATPase activity. This facilitates vesicle fusion to the target membrane followed by fusion and release of vesicle components.

Immunofluorescence, Western Blotting

Specifications

Specifications

NSF
Human NSF aa. 488-601
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG2b
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
In vitro Assay, Immunofluorescence, Western Blot
RUO
7
250μg/mL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit