Learn More
NSF Mouse anti-Bovine, Chicken, Human, Mouse, Rat, Clone: NSF-1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112435
Catalog No. 01-674-305
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A suggested positive control for this product is PC12 cells.
NSF (N-ethylmaleimide sensitive fusion protein) is a critical component of the SNARE (soluble NSF attachment protein receptors) protein complex that is involved in synaptic vesicle trafficking. Specifically, NSF has been found to be essential in membrane fusion. Furthermore, NSF has been recently demonstrated to bind other protein complexes such as AMPA receptor subunits (GluR2), GATE-16, LMA-1 and Rabs suggesting a more diverse role in the assembly of various protein complexes (Whiteheart et al., 2004).
Specifications
Specifications
NSF
Monoclonal
Unconjugated
NSF
Liquid
NSF
Recombit human NSF.
Purified
RUO
Antibody
Monoclonal
Bovine, Chicken, Human, Mouse, Rat
ELISA, Immunoprecipitation, Western Blot
NSF-1
PBS with 50% glycerol and no preservative; pH 7.5
P46459, P46460, Q9QUL6
N-ethylmaleimide sensitive fusion protein
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
18195, 419972, 4905, 504457, 60355