Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSF Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129902

Catalog No. PIPA129902


Add to cart

Description

Description

PA1-29902 detects NSF from mouse, rat samples. PA1-29902 has been successfully used in immunohistochemistry (frozen tissue), immunoprecipitation, Western blot applications. The PA1-29902 immunogen is: Synthetic peptide corresponding to amino acids around 731 of rat NSF.

NSF (N-ethylmaleimide sensitive fusion protein) is a critical component of the SNARE (soluble NSF attachment protein receptors) protein complex that is involved in synaptic vesicle trafficking. Specifically, NSF has been found to be essential in membrane
Specifications

Specifications

NSF
Polyclonal
Unconjugated
Nsf
Liquid
NSF
Synthetic peptide corresponding to amino acids around 731 of rat NSF.
Protein A
RUO
Antibody
Polyclonal
Mouse, Rat
Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
0.5 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P46460, Q9QUL6
N-ethylmaleimide sensitive fusion protein
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18195, 60355
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit