Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NSR1 Monoclonal Antibody (31C4), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA110030
Description
MA1-10030 detects Yeast Nsr1 from S. cerevisiae samples; it reacts with an epitope in the C-terminal 55% of Nsr1.
NSR1 Monoclonal antibody specifically detects NSR1 in Yeast samples. It is validated for Immunocytochemistry, Western Blot.Specifications
NSR1 | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 5mM sodium azide | |
P27476 | |
NSR1 | |
Yeast nuclear preparation. | |
RUO | |
853064 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
31C4 | |
Unconjugated | |
NSR1 | |
G7001; NSR1; Nsr1p; Nuclear localization sequence-binding protein; p67; SHE5; YGR159C | |
Mouse | |
500 μL | |
Primary | |
Yeast | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction