Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-NSUN6, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132531

Catalog No. NBP132531


Add to cart

Description

Description

NSUN6 Polyclonal antibody specifically detects NSUN6 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

NSUN6
0.84mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
NSUN6
Recombinant protein encompassing a sequence within the center region of human NSUN6. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
221078
Human
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
EC 2.1.1, EC 2.1.1.-, EC 2.1.1.52, FLJ23743, NOL1/NOP2/Sun and PUA domain-containing protein 1, NOL1/NOP2/Sun domain family 6, NOL1/NOP2/Sun domain family member 6, NOL1/NOP2/Sun domain family, member 6, NOP2/Sun domain family, member 6,4933414E04Rik, NOPD1, nucleolar protein (NOL1/NOP2/sun) and PUA domains 1, putative methyltransferase NSUN6
Rabbit
IgG
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Expected to cross react based on sequence identity: Mouse (82%), Rat (81%) Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only