Learn More
NTF3 Sheep anti-Human, Non-human primate, Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA118389
Catalog No. 01-670-641
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-18389 detects NT3 from human, rat, and chicken samples. PA1-18389 has been successfully used in immunohistochemistry, ELISA, Western blot, and inhibition assay procedures. The PA1-18389 immunogen is recombit human NT3. Reconstitute with 500 µL of distilled water.
The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintece of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
Specifications
Specifications
NTF3
Polyclonal
Unconjugated
NTF3
Lyophilized
NTF3
Recombit human NT3.
Protein G
RUO
Antibody
Polyclonal
Human, Non-human Primate, Rat
ELISA, Immunohistochemistry, Inhibition Assays, Western Blot
1 mg/mL
Antibody with no preservative
P20783, P18280
Neurotrophin-3, Neurotrophic factor, HDNF, Nerve growth factor 2, NGF-2, Ntf3, Ntf-3
Sheep
IgG
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4908, 81737