anti-Neurotrophin-3 Polyclonal, Thermo Scientific™

Neurotrophin-3 Polyclonal Ab PA1-18384 from Thermo Scientific™ detects Neurotrophin-3 in Ck, Hu and Rt samples, and is validated for ELISA, IHC, Inhibition Assays and WB

Manufacturer: Invitrogen PA118391

 View more versions of this product

Catalog No. 01-670-643

  Check Availability
Add to Cart

    Specifications

    Formulation 500μg purified antibody (1mg/mL) and no preservative
    Applications Enzyme Linked Immunosorbent Assay,Immunohistochemistry,Inhibition Assays,Western Blot
    Cross Reactivity Human,Non-human primate,Rat
    Storage Requirements Avoid Freeze/Thaw Cycles
    Gene ID (Entrez) 4908, 81737
    Gene Accession No. P18280, P20783
    Gene Alias Neurotrophin-3, Neurotrophic factor, HDNF, Nerve growth factor 2, NGF-2, Ntf3, Ntf-3
    Gene Symbols NTF3
    Species Reactivity Human, Non-human primate, Rat
    Antigen Neurotrophin-3
    Format Purified
    Host Species Rabbit
    Immunogen Recombinant human NT3.
    Isotype IgG
    Quantity 500μg
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Conjugate Unlabeled

    Neurotrophin-3 Polyclonal Ab PA1-18384 from Thermo Scientific™ detects Neurotrophin-3 in Ck, Hu and Rt samples, and is validated for ELISA, IHC, Inhibition Assays and WB

    The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
    Host Species: Rabbit
    Cross Reactivity: Human, Non-human primate, Rat
    Isotype: IgG
    Immunogen: Recombinant human NT3.
    Formulation: 500µg purified antibody (1mg/mL) and no preservative
    Synonyms: Neurotrophin-3, Neurotrophic factor, HDNF, Nerve growth factor 2, NGF-2, Ntf3, Ntf-3
    Uniprot Accession: P18280, P20783
    Entrez GeneID: 4908, 81737
    Gene Symbol: NTF3
    PA1-18391 detects NT3 from human, rat, and chicken samples. PA1-18391 has been successfully used in immunohistochemistry, ELISA, Western blot, and inhibition assay procedures. The PA1-18391 immunogen is recombinant human NT3. Reconstitute with 500 ul of distilled water.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit