Learn More
NTAL Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA119440
Catalog No. 01-676-756
Add to cart
Description
Description
This antibody recognizes the defined epitope (aa 91-243) of human Non-T cell Activation Linker (NTAL). It will not cross-react with mouse.
NTAL (Non-T cell activation linker), also known as LAB (Linker for activation of B cells), is a 30 kDa double-palmitoylated transmembrane adaptor protein expressed by B cells, NK cells, mast cells and macrophages. It is a negative regulator of early stages of BCR-dependent B cell signaling and serves as a negative regulator also in mast cells. However, in mast cells, NTAL also contributes to some activation processes, partially overlapping with LAT function.
Specifications
Specifications
NTAL
Polyclonal
PBS with 15mM sodium azide; pH 7.4
Q9GZY6
LAB
Rabbit
IgG
100 μg
4° C, do not freeze
Primary
7462
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LAT2
Liquid
LAT2
Recombit cytoplasmic domain (aa 91-243) of human NTAL.
Protein A
RUO
Antibody
Polyclonal
Human