Learn More
NTAL Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA119442
Catalog No. 01-676-758
Add to cart
Description
Description
This antibody will not cross-react with human.
NTAL (Non-T cell activation linker), also known as LAB (Linker for activation of B cells), is a 30 kDa double-palmitoylated transmembrane adaptor protein expressed by B cells, NK cells, mast cells and macrophages. It is a negative regulator of early stages of BCR-dependent B cell signaling and serves as a negative regulator also in mast cells. However, in mast cells, NTAL also contributes to some activation processes, partially overlapping with LAT function.
Specifications
Specifications
NTAL
Polyclonal
PBS with 15mM sodium azide; pH 7.4
Q9JHL0, Q8CGL2
LAB
Rabbit
IgG
100 μg
4° C, do not freeze
Primary
317676, 56743
Immunocytochemistry, Western Blot
Unconjugated
Lat2
Liquid
LAT2
Recombit mouse NTAL (aa 30-203).
Protein A
RUO
Antibody
Polyclonal
Mouse, Rat