Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

neurotrophic tyrosine kinase, receptor, type 3, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human NTRK3 protein.

Manufacturer:  Abnova Corporation H00004916B01

Catalog No. 89-005-875


Add to cart

Description

Description

This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation and may play a role in the development of proprioceptive neurons that sense body position. Mutations in this gene have been associated with medulloblastomas, secretory breast carcinomas and other cancers. [provided by RefSeq

Sequence: MDVSLCPAKCSFWRIFLLGSVWLDYVGSVLACPANCVCSKTEINCRRPDDGNLFPLLEGQDSGNSNGNASINITDISRNITSIHIENWRSLHTLNAVDMELYTGLQKLTIKNSGLRSIQPRAFAKNPHLRYINLSSNRLTTLSWQLFQTLSLRELQLEQNFFNCSCDIRWMQLWQEQGEAKLNSQNLYCINADGSQLPLFRMNISQCDLPEISVSHVNLTVREGDNAVITCNGSGSPLPDVDWIVTGLQSINTHQTNLNWTNVHAINLTLVNVTSEDNGFTLTCIAENVVGMSNASVALTVYYPPRVVSLEEPELRLEHCIEFVVRGNPPPTLHWLHNGQPLRESKIIHVEYYQEGEISEGCLLFNKPTHYNNGNYTLIAKNPLGTANQTINGHFLKEPFPESTDNFILFDEVSPTPPITVTHKPEEDTFGVSIAVGLAAFACVLLVVLFVMINKYGRRSKFGMKGPVAVISGEEDSASPLHHINHGITTPSSLDAGPDTVVIGMTRIPVIENPQYFRQGHNCHKPDTWVFSNIDNHGILNLKDNRDHLVPSTHYIYEEPEVQSGEVSYPRSHGFREIMLNPISLPGHSKPLNHGIYVEDVNVYFSKGRHGF
Specifications

Specifications

neurotrophic tyrosine kinase, receptor, type 3
Unconjugated
No additive
NM_001007156.1
TRKC|gp145(trkC)
Mouse
50 uL
Kinases
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human NTRK3 protein.
NTRK3
Serum
NTRK3
NTRK3 (NP_001007157.1, 1 a.a. ~ 612 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
4916
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit