Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUAK2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523673
Description
Specifications
NUAK2 | |
Polyclonal | |
NUAK2 | |
SNARK, NUAK family SNF1-like kinase 2, SNF1/AMP activated protein kinase, SNF1/AMP kinase-related kinase, omphalocele kinase 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
74137, 81788 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q9H093, Q8BZN4 | |
NUAK2 | |
recombit protein from human NUAK2. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction