Learn More
NUBP1 Mouse anti-Human, Clone: 2B11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519412
Catalog No. PIMA519412
Add to cart
Description
Description
Peptide Sequence: PIIGVVENMS GFICPKCKKE SQIFPPTTGG AELMCQDLEV PLLGRVPLDP LIGKNCDKGQ SFFIDAPDSP ATLAYRSIIQ RIQEFCNLHQ SKEENLISS
Component of the cytosolic iron-sulfur (Fe/S) protein assembly (CIA) machinery (PubMed:18573874). Required for maturation of extramitochondrial Fe-S proteins (PubMed:18573874). The NUBP1-NUBP2 heterotetramer forms a Fe-S scaffold complex, mediating the de novo assembly of an Fe-S cluster and its transfer to target apoproteins (PubMed:18573874). Implicated in the regulation of centrosome duplication (By similarity). Negatively regulates cilium formation and structure (By similarity).
Specifications
Specifications
NUBP1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P53384
NUBP1
NUBP1 (NP_002475.1, 222 a.a. ∽ 320 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
2B11
Unconjugated
NUBP1
Liquid
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4682