Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Nuclear Factor Erythroid Derived 2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131540

Catalog No. NBP131540


Add to cart

Description

Description

Nuclear Factor Erythroid Derived 2 Polyclonal antibody specifically detects Nuclear Factor Erythroid Derived 2 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

Nuclear Factor Erythroid Derived 2
1.03mg/mL
Western Blot 1:500-1:3000, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation
Affinity Purified
NFE2
Recombinant protein encompassing a sequence within the center region of human NF-E2. The exact sequence is proprietary.
41 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Monkey (97%). Not yet tested in any other species.
Western Blot, ChIP assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
1XPBS, 1% BSA, 20% Glycerol (pH7). with 0.025% Proclin 300
Leucine zipper protein NF-E2, NF-E2, nuclear factor (erythroid-derived 2), 45kD, nuclear factor (erythroid-derived 2), 45kDa, Nuclear factor, erythroid-derived 2 45 kDa subunit, p45, p45 NF-E2, transcription factor NF-E2 45 kDa subunit
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
4778
Human, Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only