Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UACA Mouse anti-Human, Clone: AE-5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135738

Catalog No. 01-672-044


Add to cart

Description

Description

Regulates APAF1 expression and plays an important role in the regulation of stress-induced apoptosis. Promotes apoptosis by regulating three pathways, apoptosome up-regulation, LGALS3/galectin-3 down-regulation and NF-kappa-B inactivation. Regulates the redistribution of APAF1 into the nucleus after proapoptotic stress. Down-regulates the expression of LGALS3 by inhibiting NFKB1.
Specifications

Specifications

UACA
Monoclonal
0.1 mg/mL
PBS with 0.02% sodium azide
Q9BZF9
UACA
Human UACA
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
AE-5
Unconjugated
UACA
Liquid
Mouse
IgG1, kappa
100 μg
4° C
Primary
55075
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit