Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleolin Mouse anti-Canine, Human, Monkey, Clone: 3G4B2, MilliporeSigma™

Mouse Monoclonal Antibody

Manufacturer:  MilliporeSigma 05565

Catalog No. 05-565-MI


Add to cart

Description

Description

Anti-Nucleolin Antibody, clone 3G4B2 is a high quality Mouse Monoclonal Antibody for the detection of Nucleolin and has been validated in immunoprecipitation, western blot, immunocytochemistry.
Specifications

Specifications

Nucleolin
3G4B2
mouse culture supernatant containing 0.05% sodium azide prior to the addition of glycerol to 30%
Supernatant
Murine
IgG
100μL
Primary
Canine, Human, Monkey
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_005381
NCL, FLJ45706, nucleolin, C23, Nucleolin
Nuclear matrix extract from MDCK cells
Unpurified
−20°C for 24 months.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only