Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Nucleoporin 107, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP149841

Catalog No. NBP149841


Add to cart

Description

Description

Nucleoporin 107 Polyclonal antibody specifically detects Nucleoporin 107 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

Nucleoporin 107
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000
NP_065134.1
nuclear pore complex protein Nup107, nucleoporin 107kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin
Goat
IgG
0.1 mg
Core ESC Like Genes, Stem Cell Markers
Primary
57122
Human, Mouse
ELISA, Western Blot
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
NUP107
Peptide with sequence C-PLPAEDDNAIREH corresponding to internal region according to NP_065134.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody recognizes and epitope within the internal region.This antibody is specific to the internal region. Expected from sequence similarity: Mouse, Rat, Dog and Cow.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only