Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleoporin p62 Mouse, FITC, Clone: 53, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611962

Catalog No. BDB611962


Add to cart

Description

Description

The nuclear pore complex (NPC) acts as a gate to mediate active transport of proteins and RNA into and out of the nucleus. Proteins actively transported into the nucleus through the NPC require specific nuclear localization sequences. Many of these nucleoporins contain N-acetylglucosamine (GlcNAc) residues that are O-linked to serine or threonine. p62 is the best characterized member of a group of nucleoporins that line the central region of the NPC. A tightly associated complex is formed by p62 and two other nucleoporins, p54 and p58. p54 binds to a carboxy-terminal coiled-coil domain of p62 and p58 binds to a dimer of p54. The amino-terminal domain of p62 contains a series of XFXFX repeats and is joined to the coiled-coil domain by a threonine-rich linker segment. The major role of p62 is maintenance of the structural integrity of NPCs.

Specifications

Specifications

Nucleoporin p62
53
FITC
Affinity Purified
Human Nucleoporin aa. 24-178
50μg
Cell Biology
Primary
Chicken, Human, Murine, Rat
Immunofluorescence
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit