Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleostemin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511440

Catalog No. PIPA511440

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GNL3 is found in the nucleoli of embryonic stem cells, adult CNS stem cells, primitive cells in the bone marrow and cancer cells. It is not in the differentiated cells of most adult tissues. It has been suggested to play a role in controlling the cell-cycle progression in stem cells and cancer cells, and may be required to maintain the proliferative capacity of stem cells.
Specifications

Specifications

Nucleostemin
Polyclonal
PBS with 0.09% sodium azide
Q9BVP2
Guanine nucleotide-binding protein-like 3; Nucleolar GTP-binding protein 3; Nucleostemin; E2-induced gene 3 protein; Novel nucleolar protein 47; NNP47; E2IG3, NS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26354
Immunohistochemistry, Western Blot
Unconjugated
GNL3
Liquid
GNL3
KLH conjugated synthetic peptide between 458-487 amino acids from the C-terminal region of human Nucleostemin (GNL3)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit