Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT2 Monoclonal Antibody (S1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA518404
Description
Peptide Sequence: MALRACGLII FRRCLIPKVD NNAIEFLLLQ ASDGIHHWTP PKGHVEPGED DLETALRETQ EEAGIEAGQL TIIEGFKREL NYVARNKPKT VIYWLAEVKD YDVEIRLSHE HQAYRWLGLE EACQLAQFKE MKAALQEGHQ FLCSIEA
NUDT2 Monoclonal antibody specifically detects NUDT2 in Human samples. It is validated for ELISA, Western Blot.Specifications
NUDT2 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P50583 | |
NUDT2 | |
NUDT2 (AAH04926, 1 a.a. ∽ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
S1 | |
Unconjugated | |
NUDT2 | |
2310051L06Rik; AA939917; Ap4A hydrolase; Ap4A hydrolase 1; Ap4Aase; APAH1; bis(5'-nucleosyl)-tetraphosphatase; bis(5'-nucleosyl)-tetraphosphatase (asymmetrical); bis(5'-nucleosyl)-tetraphosphatase [asymmetrical]; Diadenosine 5',5'''-P1,P4-tetraphosphate asymmetrical hydrolase; diadenosine 5',5''-P1,P4-tetraphosphate pyrophosphohydrolase; diadenosine tetraphosphatase; MGC10404; nucleoside diphosphate-linked moiety X motif 2; nudix (nucleoside diphosphate linked moiety X)-type motif 2; nudix hydrolase 2; nudix motif 2; Nudt2 | |
Mouse | |
Affinity chromatography | |
RUO | |
318 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction