Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NULP1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527618

Catalog No. PIPA527618


Add to cart

Description

Description

PA5-27618 targets NULP1 in WB applications and shows reactivity with mouse samples. The PA5-27618 immunogen is recombit fragment corresponding to a region within amino acids 1 and 266 of Human NULP1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

TCF25 is a member of the basic helix-loop-helix (bHLH) family of transcription factors that are important in embryonic development (Steen and Lindholm, 2008 [PubMed 18068114]).
Specifications

Specifications

NULP1
Polyclonal
Unconjugated
Tcf25
Liquid
TCF25
Recombit fragment corresponding to a region within amino acids 1 and 266 of Human NULP1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q8R3L2
Hulp1, NULP1, PRO2620, hKIAA1049, NULP1, TCF-25, nuclear localized protein 1, transcription factor 25
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
66855
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit