Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NuMA Mouse anti-Human, Clone: A73-B/D12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135739

Catalog No. 01-672-045


Add to cart

Description

Description

NuMA (nuclear mitotic apparatus) is a long coil-coiled protein that plays a role in the interphase and mitosis phases of a cell cycle. In interphase, it has been hypothesized to play a structural role in the nucleoskeleton that may be important in nuclear organization and functions. During mitosis, it is associated with spindle microtubule organization and chromosome positioning. Therefore, this antibody is useful as a cell cycle marker.

Specifications

Specifications

NuMa
Monoclonal
0.1 mg/mL
PBS with 0.05% sodium azide
Q14980
NUMA
Mouse
IgM
100 μg
4° C
Primary
4926
ELISA, Immunohistochemistry (Frozen), Western Blot
A73-B/D12
Unconjugated
NUMA1
Liquid
NUMA1
Colon carcinoma cell line Ls-174T.
Purified
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit