Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

NUMB Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132452

Catalog No. PIPA132452



Description

Description

PA1-32452 detects Numb from mouse, human samples. PA1-32452 has been successfully used in Western blot applications. The PA1-32452 immunogen is: Synthetic peptide corresponding to a C-terminal region of the mouse numb protein sequence (between residues 600-653).

The protein encoded by this gene plays a role in the determination of cell fates during development. The encoded protein, whose degradation is induced in a proteasome-dependent manner by MDM2, is a membrane-bound protein that has been shown to associate with EPS15, LNX1, and NOTCH1. Four transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

NUMB
Polyclonal
Unconjugated
NUMB
Liquid
NUMB
Synthetic peptide corresponding to a C-terminal region of the murine numb protein sequence (between residues 600-653).
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
3.5 mg/mL
tris glycine with 0.15M NaCl and 0.05% sodium azide
P49757, Q9QZS3, Q2LC84
S171
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18222, 29419, 8650
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit