Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gp210 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526961

Catalog No. PIPA526961

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

NUP210 nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex.
Specifications

Specifications

gp210
Polyclonal
PBS with 0.09% sodium azide
Q8TEM1
GP210, POM210, nuclear envelope pore membrane protein POM 210, nuclear pore membrane glycoprotein 210, nuclear pore protein gp210, nucleoporin Nup210, pore membrane protein of 210 kDa
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23225
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
NUP210
Liquid
NUP210
KLH conjugated synthetic peptide between 202-231 amino acids from the N-terminal region of human NUP210
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit