Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUPL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526571

Catalog No. PIPA526571


Add to cart

Description

Description

Required for the export of mRNAs containing poly(A) tails from the nucleus into the cytoplasm. In case of infection by HIV-1, it may participate in the docking of viral Vpr at the nuclear envelope.
Specifications

Specifications

NUPL2
Polyclonal
PBS with 0.09% sodium azide
O15504
CG1, NLP-1, NLP_1, hCG1, H_RG271G13.9, NUP42 homolog, nucleoporin hCG1, nucleoporin-like protein 1, nucleoporin-like protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11097
Western Blot
Unconjugated
NUPL2
Liquid
NUPL2
KLH conjugated synthetic peptide between 88-115 amino acids from the N-terminal region of human NUPL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit