Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUSAP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524479

Catalog No. PIPA524479

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

NUSAP1 is a nucleolar-spindle-associated protein that plays a role in spindle microtubule organization.
Specifications

Specifications

NUSAP1
Polyclonal
PBS with 0.09% sodium azide
Q9BXS6
ANKT, BM037, LNP, NUSAP, PRO0310p1, Q0310, SAPL, nucleolar and spindle-associated protein 1, nucleolar protein ANKT
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51203
Immunohistochemistry, Western Blot
Unconjugated
NUSAP1
Liquid
NUSAP1
KLH conjugated synthetic peptide between 27-56 amino acids from the N-terminal region of human NUSAP
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit