Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NXNL2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524065

Catalog No. PIPA524065

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The specific function of NXNL2 remains unknown.
Specifications

Specifications

NXNL2
Polyclonal
PBS with 0.09% sodium azide
Q5VZ03, Q9D531
C9orf121, RDCVF2, nucleoredoxin-like protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
158046, 75124
Immunohistochemistry, Western Blot
Unconjugated
NXNL2
Liquid
NXNL2
KLH conjugated synthetic peptide between 80-108 amino acids from the central region of human NXNL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit