Learn More
NXPH3 Mouse anti-Human, Clone: 4C8, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA521314
Catalog No. PIMA521314
Add to cart
Description
Description
Peptide Sequence: MQLTRCCFVF LVQGSLYLVI CGQDDGPPGS EDPERDDHEG QPRPRVPRKR GHISPKSRPM ANSTLLGLLA PPGEAWGILG QPPNRPNHSP PPSAKVKKIF GWGDFYSNIK TVALNLLVTG KIVDHGNGTF SVHFQHNATG QGNISISLVP PSKAVEFHQE QQIFIEAKAS KIFNCRMGWE KVERGRRTSL CTHDPAKICS RDHAQSSATW SCSQPFKVVC VYIAFYSTDY RLVQKVCPDY NYHSDTPYYP SG
May be signaling molecules that resemble neuropeptides. Ligand for alpha-neurexins (By similarity).
Specifications
Specifications
NXPH3
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
O95157
KIAA1159, NPH3
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
11248
ELISA, Western Blot
4C8
Unconjugated
NXPH3
Liquid
NXPH3
NXPH3 (AAH22541, 1 a.a. ∽ 252 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human