Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
O-GlcNAc Mouse anti-Human, Clone: 18B10.C7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051244
Description
Anti-O-GlcNAc Antibody, clone 18B10.C7(3) is an antibody against O-GlcNAc for use in western blotting.Specifications
O-GlcNAc | |
Monoclonal | |
Unconjugated | |
NP_858058.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Western Blot | |
18B10.C7 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
OGT, HRNT1, O-GLCNAC | |
SANM-conjugated linear peptide corresponding to human O-GluNAc. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only