Learn More
OAS3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA513211
Catalog No. PIPA513211
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
OAS3 is an interferon inducible protein that belongs to the 2-5A synthetase family, may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. OAS3 synthesizes preferentially dimeric 2′,5′-oligoadenylate molecules. GTP can be an alternative substrate. OAS3 binds double-stranded RNA and polymerizes ATP into PPP(A2′P5′A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. The protein is present at high level in placenta trophoblast.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OAS3
Polyclonal
OAS3
OAS3
KLH conjugated synthetic peptide between 1056-1087 amino acids from the C-terminal region of human OAS3
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
Q9Y6K5
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
4940
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Recommendations
Recommendations