Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB3L1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513212

Catalog No. PIPA513212

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

OASIS is a putative CREB/ATF family transcription factor with a putative C-terminal hydrophobic transmembrane domain. It can activate transcription through box-B elements but not through CRE of somatostatin. Shortening of the OASIS transmembrane domain results in a significant increase in transcriptional activity and changes its subcellular localization from the endoplasmic reticulum to the nucleus.
Specifications

Specifications

CREB3L1
Polyclonal
PBS with 0.09% sodium azide
Q96BA8
CREB3L1
KLH conjugated synthetic peptide between 491-519 amino acids from the C-terminal region of human OASIS
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
CREB3L1
Liquid
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
90993
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit