Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ghrelin Chicken anti-Mouse, Rat, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA19597

Catalog No. 01-674-471


Add to cart

Description

Description

PA1-9597 detects Obestatin in human, mouse and rat samples. PA1-9597 has been successfully used in ELISA procedures. PA1-9597 immunogen mouse obestatin peptide (FNAPFDVGIKLSGAQYQQHGRAL), aa 76-98 of Ghrelin precursor.

GHRL encodes ghrelin-obestatin preproprotein, which generates ghrelin and obestatin. Ghrelin is an the ligand for the growth hormone secretagogue receptor type 1 and is involved in regulating growth hormone release. Ghrelin has an appeptite-stimulating effect and induces adiposity and stimulates gastric acid secretion. Obestatin may be the ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and can have an appetite-reducing effect resulting in decreased food intake.
Specifications

Specifications

Ghrelin
Polyclonal
Unconjugated
Ghrl
Liquid
GHRL
Mouse Obestatin peptide (FNAPFDVGIKLSGAQYQQHGRAL), aa 76-98 of Ghrelin precursor.
Affinity chromatography
RUO
Antibody
Polyclonal
Mouse, Rat
ELISA
1 mg/mL
PBS with 0.02% sodium azide
Q9EQX0, Q9QYH7
MTLRP, obestatin
Chicken
IgY
200 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
58991, 59301
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit